Product Details

SNP ID
rs146839918
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:65514840 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACTCACTCAGTCTTGCATGTTGA[C/G]GGTTCCAGCCGCTGCTCTAGCACAG
Phenotype
MIM: 300964 MIM: 300889
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LAS1L PubMed Links

Gene Details

Gene
LAS1L
Gene Name
LAS1 like, ribosome biogenesis factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170649.1 2083 Silent Mutation CCC,CCG P670P NP_001164120.1
NM_001170650.1 2083 Silent Mutation CCC,CCG P628P NP_001164121.1
NM_031206.4 2083 Silent Mutation CCC,CCG P687P NP_112483.1
XM_005262301.1 2083 Silent Mutation CCC,CCG P686P XP_005262358.1
XM_005262304.1 2083 Intron XP_005262361.1
XM_005262305.3 2083 Intron XP_005262362.1
XM_005262306.3 2083 Intron XP_005262363.1
XM_011531045.1 2083 Silent Mutation CCC,CCG P645P XP_011529347.1
XM_011531046.1 2083 Intron XP_011529348.1
XM_017029877.1 2083 Silent Mutation CCC,CCG P669P XP_016885366.1
XM_017029878.1 2083 Intron XP_016885367.1
XM_017029879.1 2083 Silent Mutation CCC,CCG P385P XP_016885368.1
XM_017029880.1 2083 Silent Mutation CCC,CCG P368P XP_016885369.1
Gene
ZC3H12B
Gene Name
zinc finger CCCH-type containing 12B
There are no transcripts associated with this gene.

View Full Product Details