Product Details

SNP ID
rs147816954
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154460321 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCACCGGGTGACTGGGGAGGTGTT[C/T]GTGGGCGCAGTGAACCGAGTCTTTA
Phenotype
MIM: 300453 MIM: 300022
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM50A PubMed Links

Gene Details

Gene
FAM50A
Gene Name
family with sequence similarity 50 member A
There are no transcripts associated with this gene.

Gene
MIR6858
Gene Name
microRNA 6858
There are no transcripts associated with this gene.

Gene
PLXNA3
Gene Name
plexin A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017514.4 313 Silent Mutation TTC,TTT F46F NP_059984.3
XM_005274705.4 313 Silent Mutation TTC,TTT F46F XP_005274762.2
XM_005274706.4 313 Intron XP_005274763.2
XM_006724829.3 313 Silent Mutation TTC,TTT F46F XP_006724892.1
XM_011531183.2 313 Intron XP_011529485.1

View Full Product Details