Product Details

SNP ID
rs147850709
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:57119599 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTCCTCTAAGCCACAAGCTCTC[C/T]GATAGCTGTTCCTTCTTTTCATGCC
Phenotype
MIM: 300654 MIM: 300517
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAAH2 PubMed Links

Gene Details

Gene
FAAH2
Gene Name
fatty acid amide hydrolase 2
There are no transcripts associated with this gene.

Gene
SPIN2B
Gene Name
spindlin family member 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006681.1 Intron NP_001006682.1
NM_001006682.1 Intron NP_001006683.1
NM_001006683.1 Intron NP_001006684.1
NM_001282461.1 Intron NP_001269390.1
NM_001282462.1 Intron NP_001269391.1
XM_005262008.4 Intron XP_005262065.2
XM_005262009.4 Intron XP_005262066.2
XM_005262010.1 Intron XP_005262067.1
XM_011530788.2 Intron XP_011529090.1
XM_011530789.2 Intron XP_011529091.1
XM_017029552.1 Intron XP_016885041.1

View Full Product Details