Product Details

SNP ID
rs147875150
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:69505758 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATCTTTGCAGAGACTTTTCGAAC[C/T]GACTACTCCGGCCCCCCCTCTGCGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM155B PubMed Links

Gene Details

Gene
FAM155B
Gene Name
family with sequence similarity 155 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015686.2 524 Missense Mutation CCG,CTG P159L NP_056501.2
XM_011530908.2 524 Missense Mutation CCG,CTG P159L XP_011529210.1
XM_011530909.2 524 Missense Mutation CCG,CTG P159L XP_011529211.1

View Full Product Details