Product Details

SNP ID
rs148184503
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:65081315 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACATCAAATGTAAATGGACTAAA[C/T]TCCTGAATCAAAAGATGTAGAGAGG
Phenotype
MIM: 300889
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZC3H12B PubMed Links

Gene Details

Gene
ZC3H12B
Gene Name
zinc finger CCCH-type containing 12B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010888.3 Intron NP_001010888.3
XM_011530939.2 Intron XP_011529241.1
XM_017029478.1 Intron XP_016884967.1
XM_017029479.1 Intron XP_016884968.1
XM_017029480.1 Intron XP_016884969.1
XM_017029481.1 Intron XP_016884970.1
XM_017029482.1 Intron XP_016884971.1
XM_017029483.1 Intron XP_016884972.1
XM_017029484.1 Intron XP_016884973.1

View Full Product Details