Product Details

SNP ID
rs148339434
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:72197463 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGTGGGATGGATAAGCCTGTGTT[C/T]ACAGACCCACCGGTTAAGACAAAAT
Phenotype
MIM: 300687 MIM: 300252
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ERCC6L PubMed Links

Gene Details

Gene
ERCC6L
Gene Name
ERCC excision repair 6 like, spindle assembly checkpoint helicase
There are no transcripts associated with this gene.

Gene
PIN4
Gene Name
peptidylprolyl cis/trans isomerase, NIMA-interacting 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170747.1 443 Intron NP_001164218.1
NM_006223.3 443 Silent Mutation TTC,TTT F136F NP_006214.2

View Full Product Details