Product Details

SNP ID
rs148567981
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:137567523 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGACCTTCAGCACCATGCATGAG[C/T]TGGTGACACATGTCACCATGGAGCA
Phenotype
MIM: 300265
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZIC3 PubMed Links

Gene Details

Gene
ZIC3
Gene Name
Zic family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003413.3 1337 Silent Mutation CTG,TTG L278L NP_003404.1
XM_017029802.1 1337 Silent Mutation CTG,TTG L278L XP_016885291.1

View Full Product Details