Product Details

SNP ID
rs148653835
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:70269878 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGGGGAAACGGGACTTCGTGGA[C/T]CATGTGGACACGGTGGAACCCATTG
Phenotype
MIM: 301770
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARR3 PubMed Links

Gene Details

Gene
ARR3
Gene Name
arrestin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004312.2 75 Silent Mutation GAC,GAT D25D NP_004303.2
XM_017029518.1 75 Silent Mutation GAC,GAT D25D XP_016885007.1

View Full Product Details