Product Details

SNP ID
rs149136745
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:78138051 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGCATCAACATTAGGTTTCTTAG[C/T]ATGGCTCGAATAAATTTTTGCATCA
Phenotype
MIM: 300754
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAF9B PubMed Links

Gene Details

Gene
TAF9B
Gene Name
TATA-box binding protein associated factor 9b
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015975.4 Intron NP_057059.2

View Full Product Details