Product Details

SNP ID
rs149400555
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:133217740 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTCTCCCTGGAGATGATATTCA[C/T]GGCCATCAGCACGTTTAAGGCATCG
Phenotype
MIM: 300772
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TFDP3 PubMed Links

Gene Details

Gene
TFDP3
Gene Name
transcription factor Dp family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016521.2 Intron NP_057605.3

View Full Product Details