Product Details

SNP ID
rs149552852
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:139956001 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCACTATAATATTTCATCGTATC[A/T]CTGTCATTCACATGATCACCGTATG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CXorf66 PubMed Links

Gene Details

Gene
CXorf66
Gene Name
chromosome X open reading frame 66
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013403.2 1003 Missense Mutation AGA,AGT R327S NP_001013421.1

View Full Product Details