Product Details

SNP ID
rs149830892
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:81202706 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTGAGTGGAGGCCTACCGAGGCA[C/T]TGTAATTATTCTGGGTGAGTGAGAC
Phenotype
MIM: 300385 MIM: 300190
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HMGN5 PubMed Links

Gene Details

Gene
HMGN5
Gene Name
high mobility group nucleosome binding domain 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030763.2 Intron NP_110390.1
XM_006724688.3 Intron XP_006724751.1
XM_017029839.1 Intron XP_016885328.1
XM_017029840.1 Intron XP_016885329.1
Gene
SH3BGRL
Gene Name
SH3 domain binding glutamate rich protein like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003022.2 Intron NP_003013.1
XM_011531013.1 Intron XP_011529315.1
XM_011531014.1 Intron XP_011529316.1

View Full Product Details