Product Details

SNP ID
rs150316545
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:153444915 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGTAGCGGTGGAAGAGGCTGCC[A/G]AGGCTGTAACCCTGGCGGCCCCGGG
Phenotype
MIM: 300540 MIM: 300370
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HAUS7 PubMed Links

Gene Details

Gene
HAUS7
Gene Name
HAUS augmin like complex subunit 7
There are no transcripts associated with this gene.

Gene
TREX2
Gene Name
three prime repair exonuclease 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080701.3 1060 Silent Mutation CTC,CTT L172L NP_542432.2

View Full Product Details