Product Details

SNP ID
rs150497669
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:119936407 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTTCTTGGACCTTTCTTTTGAA[C/T]GGCTAGACTTCTTCTTTTTTTCTTC
Phenotype
MIM: 300766
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NKAP PubMed Links

Gene Details

Gene
NKAP
Gene Name
NFKB activating protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024528.3 730 Missense Mutation CAT,CGT H188R NP_078804.2
XM_017029842.1 730 Missense Mutation CAT,CGT H89R XP_016885331.1

View Full Product Details