Product Details

SNP ID
rs150547120
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:96884747 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAGCCTTGAGGTATTGAAAATTC[A/G]TGTCCTAGAGGACATGAACGAGTTC
Phenotype
MIM: 300108 MIM: 300767
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DIAPH2 PubMed Links

Gene Details

Gene
DIAPH2
Gene Name
diaphanous related formin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006729.4 840 Intron NP_006720.1
NM_007309.3 840 Intron NP_009293.1
Gene
RPA4
Gene Name
replication protein A4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013347.4 840 Missense Mutation CAT,CGT H146R NP_037479.1

View Full Product Details