Product Details

SNP ID
rs150937949
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:130404888 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTGGATGTTGGGAAGTTCACAG[C/T]AGGTGCAGCCTATAATTTCTACCCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RBMX2 PubMed Links

Gene Details

Gene
RBMX2
Gene Name
RNA binding motif protein, X-linked 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016024.2 Intron NP_057108.2

View Full Product Details