Product Details
- SNP ID
-
rs151197516
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:64268575 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGTAGTCTGCCTCCTTGGTGCTGG[A/C]CTTGGAGATGCCTGTGTCCCCCAAG
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MTMR8
PubMed Links
Gene Details
- Gene
- MTMR8
- Gene Name
- myotubularin related protein 8
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_017677.3 |
2167 |
Missense Mutation |
GCC,TCC |
A693S |
NP_060147.2 |
View Full Product Details