Product Details

SNP ID
rs1059529
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:77721377 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGAGGATGGTGTCCTTGACGGC[T/A]GCAAAGACAAAGCGGATATTCTCGG
Phenotype
MIM: 600998
Polymorphism
T/A, Transversion substitution
Allele Nomenclature
Literature Links
GNAQ PubMed Links

Gene Details

Gene
GNAQ
Gene Name
G protein subunit alpha q
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002072.4 1240 Silent Mutation GCA,GCT A342A NP_002063.2
XM_017014628.1 1240 Silent Mutation GCA,GCT A284A XP_016870117.1

View Full Product Details