Product Details

SNP ID
rs1062873
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:73640953 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACACACAGAAAGAGATCACAGAC[T/G]CCCTACCTTAGAAGAAGGGAGGTGG
Phenotype
MIM: 606716
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
ALMS1P1 PubMed Links

Gene Details

Gene
ALMS1P1
Gene Name
ALMS1, centrosome and basal body associated protein pseudogene 1
There are no transcripts associated with this gene.

Gene
NAT8
Gene Name
N-acetyltransferase 8 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003960.3 843 Missense Mutation AGT,CGT S226R NP_003951.3

View Full Product Details