Product Details

SNP ID
hCV175675747
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:13306406 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCAAAGCACACTTACATTTTCCA[G/A]AAACTTTGTTATCTTCTCTGCACTG
Phenotype
MIM: 612655
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
TBC1D7 PubMed Links

Gene Details

Gene
TBC1D7
Gene Name
TBC1 domain family member 7
There are no transcripts associated with this gene.

Gene
TBC1D7-LOC100130357
Gene Name
TBC1D7-LOC100130357 readthrough
There are no transcripts associated with this gene.

View Full Product Details