Product Details

SNP ID
rs11544840
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:134478669 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACTTCATTTCTTGTTCTTTGACA[A/G]ATTCTGTGGTGCCACCATTGTCTTG
Phenotype
MIM: 614484 MIM: 614724
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANAPC13 PubMed Links

Gene Details

Gene
ANAPC13
Gene Name
anaphase promoting complex subunit 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242374.1 877 Missense Mutation TCT,TTT S49F NP_001229303.1
NM_001242375.1 877 Missense Mutation TCT,TTT S49F NP_001229304.1
NM_015391.3 877 Intron NP_056206.1
Gene
CEP63
Gene Name
centrosomal protein 63
There are no transcripts associated with this gene.

View Full Product Details