Product Details

SNP ID
rs11548299
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30489922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTATTGGGACCGGGAGACACGGAG[C/G]GCCAGGGACACCGCACAGATTTTCC
Phenotype
MIM: 143010
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HLA-E PubMed Links

Gene Details

Gene
HLA-E
Gene Name
major histocompatibility complex, class I, E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005516.5 399 Silent Mutation AGC,AGG S87R NP_005507.3
XM_017010807.1 399 Missense Mutation AGC,AGG S128R XP_016866296.1
XM_017010808.1 399 Silent Mutation AGC,AGG S128R XP_016866297.1
XM_017010809.1 399 Missense Mutation AGC,AGG S87R XP_016866298.1

View Full Product Details