Product Details
- SNP ID
-
hCV175676954
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:73769962 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGCTGCCGATGAAGGCCGACACGC[G/T]CCACATGGGCAACGCGCAGCACACG
- Phenotype
-
MIM: 602910
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CLDN3
PubMed Links
Gene Details
- Gene
- CLDN3
- Gene Name
- claudin 3
There are no transcripts associated with this gene.
View Full Product Details