Product Details

SNP ID
rs11555801
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:110244212 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTCCTTATTGGCTCCAGAAAAT[T/C]CACCCACCTGTTAAGAGAATATTGA
Phenotype
MIM: 187700
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
TXN PubMed Links

Gene Details

Gene
TXN
Gene Name
thioredoxin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244938.1 468 Missense Mutation GAA,GGA E68G NP_001231867.1
NM_003329.3 468 Missense Mutation GAA,GGA E88G NP_003320.2

View Full Product Details