Product Details

SNP ID
rs1264457
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30490287 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATGGCTGCGAGCTGGGGCCCGAC[G/A]GGCGCTTCCTCCGCGGGTATGAACA
Phenotype
MIM: 143010
Polymorphism
G/A, Transition Substitution
Allele Nomenclature
Literature Links
HLA-E PubMed Links

Gene Details

Gene
HLA-E
Gene Name
major histocompatibility complex, class I, E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005516.5 520 Missense Mutation AGG,GGG R128G NP_005507.3
XM_017010807.1 520 Missense Mutation AGG,GGG R169G XP_016866296.1
XM_017010808.1 520 Missense Mutation AGG,GGG R169G XP_016866297.1
XM_017010809.1 520 Missense Mutation AGG,GGG R128G XP_016866298.1

View Full Product Details