Product Details

SNP ID
rs179468
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:23767587 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGCGGGTGGGCAGAGGGCGCGGC[G/T]GGGCGGCTGCGGGGGTGGGAGGAAG
Phenotype
MIM: 615903 MIM: 185261
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C22orf15 PubMed Links

Gene Details

Gene
C22orf15
Gene Name
chromosome 22 open reading frame 15
There are no transcripts associated with this gene.

Gene
CHCHD10
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301339.1 Intron NP_001288268.1
NM_213720.2 Intron NP_998885.1
Gene
MMP11
Gene Name
matrix metallopeptidase 11
There are no transcripts associated with this gene.

View Full Product Details