Product Details

SNP ID
rs7187744
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29780416 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTACCCCAGTAGCTGCAGCAGATG[C/T]TGAGCCTCCTCTCCTTGGCAGGGGC
Phenotype
MIM: 603213
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIF22 PubMed Links

Gene Details

Gene
KIF22
Gene Name
kinesin family member 22
There are no transcripts associated with this gene.

Gene
ZG16
Gene Name
zymogen granule protein 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152338.3 654 Silent Mutation TGC,TGT C167C NP_689551.2
XM_011545921.2 654 Silent Mutation TGC,TGT C167C XP_011544223.1

View Full Product Details