Product Details
- SNP ID
-
rs936212
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:40289342 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCCTCCAGCTTCTCCTGGTGCCTC[C/T]CCAAGTTCTCCGTCATCTGGGTGGG
- Phenotype
-
MIM: 604114
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ANKRD63
PubMed Links
Gene Details
- Gene
- ANKRD63
- Gene Name
- ankyrin repeat domain 63
There are no transcripts associated with this gene.
- Gene
- PLCB2
- Gene Name
- phospholipase C beta 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001284297.1 |
3596 |
Missense Mutation |
GAG,GGG |
E1091G |
NP_001271226.1 |
NM_001284298.1 |
3596 |
Missense Mutation |
GAG,GGG |
E1080G |
NP_001271227.1 |
NM_001284299.1 |
3596 |
Intron |
|
|
NP_001271228.1 |
NM_004573.2 |
3596 |
Missense Mutation |
GAG,GGG |
E1095G |
NP_004564.2 |
XM_011521674.2 |
3596 |
Missense Mutation |
GAG,GGG |
E1095G |
XP_011519976.1 |
XM_017022314.1 |
3596 |
Missense Mutation |
GAG,GGG |
E1108G |
XP_016877803.1 |
XM_017022315.1 |
3596 |
Missense Mutation |
GAG,GGG |
E1107G |
XP_016877804.1 |
XM_017022316.1 |
3596 |
Missense Mutation |
GAG,GGG |
E1104G |
XP_016877805.1 |
XM_017022317.1 |
3596 |
Missense Mutation |
GAG,GGG |
E1108G |
XP_016877806.1 |
XM_017022318.1 |
3596 |
Missense Mutation |
GAG,GGG |
E1093G |
XP_016877807.1 |
XM_017022319.1 |
3596 |
Missense Mutation |
GAG,GGG |
E1108G |
XP_016877808.1 |
View Full Product Details