Product Details

SNP ID
rs1049880
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:177596041 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTTGGGGTGGGGCCTGCAGAGT[C/T]TTAGTTACTGATTTCATTTTCAATA
Phenotype
MIM: 604327
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
B4GALT7 PubMed Links

Gene Details

Gene
B4GALT7
Gene Name
beta-1,4-galactosyltransferase 7
There are no transcripts associated with this gene.

Gene
TMED9
Gene Name
transmembrane p24 trafficking protein 9
There are no transcripts associated with this gene.

View Full Product Details