Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001098815.2 | 7402 | UTR 3 | NP_001092285.1 | ||
NM_001136030.2 | 7402 | UTR 3 | NP_001129502.1 | ||
NM_001261844.1 | 7402 | UTR 3 | NP_001248773.1 | ||
NM_014796.2 | 7402 | UTR 3 | NP_055611.1 | ||
XM_005269247.2 | 7402 | UTR 3 | XP_005269304.1 | ||
XM_006719715.3 | 7402 | UTR 3 | XP_006719778.1 | ||
XM_011539035.2 | 7402 | UTR 3 | XP_011537337.1 | ||
XM_011539036.1 | 7402 | UTR 3 | XP_011537338.1 | ||
XM_011539037.2 | 7402 | UTR 3 | XP_011537339.1 | ||
XM_017020262.1 | 7402 | UTR 3 | XP_016875751.1 | ||
XM_017020263.1 | 7402 | UTR 3 | XP_016875752.1 |