Product Details

SNP ID
rs11253042
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:5204621 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGCAGAAGAGTCCTAAGTATAATG[A/C]CTAAGCCATTTTCTTGAAGAGTACA
Phenotype
MIM: 600451
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AKR1C4 PubMed Links

Gene Details

Gene
AKR1C4
Gene Name
aldo-keto reductase family 1, member C4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001818.3 Intron NP_001809.3

View Full Product Details