Product Details

SNP ID
rs11466819
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:157478680 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAAGCATTAGTAGAACTGGTTGCC[A/G]AAAGTCTATCAAATTCCAAAACATT
Phenotype
MIM: 603640 MIM: 609383
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAM19 PubMed Links

Gene Details

Gene
ADAM19
Gene Name
ADAM metallopeptidase domain 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033274.4 4370 UTR 3 NP_150377.1
XM_005266003.2 4370 Intron XP_005266060.1
XM_011534682.2 4370 UTR 3 XP_011532984.1
XM_017010009.1 4370 UTR 3 XP_016865498.1
Gene
NIPAL4
Gene Name
NIPA like domain containing 4
There are no transcripts associated with this gene.

View Full Product Details