Product Details
- SNP ID
-
rs11546416
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:56106408 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGAGGTGCGGCTTCGGCTTCCACG[T/C]GGCATCCTAGGTAAGCGACGGCTAG
- Phenotype
-
MIM: 616244
- Polymorphism
- T/C, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CHCHD2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10043] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CHCHD2
- Gene Name
- coiled-coil-helix-coiled-coil-helix domain containing 2
- Gene
- NUPR2
- Gene Name
- nuclear protein 2, transcriptional regulator
There are no transcripts associated with this gene.
View Full Product Details