Product Details

SNP ID
rs11550419
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:216038534 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAAATAAGTCATTGTATAATTCT[A/C]ATTTTAATAATCTTAATTACGACAC
Phenotype
MIM: 605843
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PECR PubMed Links

Gene Details

Gene
PECR
Gene Name
peroxisomal trans-2-enoyl-CoA reductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018441.5 1728 UTR 3 NP_060911.2
XM_011511478.2 1728 UTR 3 XP_011509780.1

View Full Product Details