Product Details

SNP ID
rs11547687
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:154207009 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCACCAGTTCCTGTCTGCTTTTTC[A/C]ATCAATCAACTCTAAGATGCCCTAA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C1orf189 PubMed Links

Gene Details

Gene
C1orf189
Gene Name
chromosome 1 open reading frame 189
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010979.2 1483 Intron NP_001010979.1
Gene
C1orf43
Gene Name
chromosome 1 open reading frame 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098616.2 1483 UTR 3 NP_001092086.1
NM_001297717.1 1483 UTR 3 NP_001284646.1
NM_001297718.1 1483 UTR 3 NP_001284647.1
NM_001297720.1 1483 UTR 3 NP_001284649.1
NM_001297721.1 1483 Intron NP_001284650.1
NM_001297723.1 1483 Intron NP_001284652.1
NM_015449.3 1483 UTR 3 NP_056264.1
NM_138740.3 1483 UTR 3 NP_620077.1

View Full Product Details