Product Details

SNP ID
rs11556871
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:70987895 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCTTGGGGAAAGAGATGGAAGAGA[A/G]TGCATCAGAGAGGCTGCCTATAGCC
Phenotype
MIM: 617009
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD53 PubMed Links

Gene Details

Gene
ANKRD53
Gene Name
ankyrin repeat domain 53
There are no transcripts associated with this gene.

Gene
TEX261
Gene Name
testis expressed 261
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144582.2 1410 UTR 3 NP_653183.2

View Full Product Details