Product Details
- SNP ID
-
rs11557532
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:82444072 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAGACAGCTCTGTGGGGCTCTCAA[G/A]GCAGTGCAGCTCCAGGAAGCTGGTG
- Phenotype
-
MIM: 616864
- Polymorphism
- G/A, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C17orf62
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2306758] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C17orf62
- Gene Name
- chromosome 17 open reading frame 62
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001033046.3 |
661 |
Missense Mutation |
CTT,TTT |
L166F |
NP_001028218.1 |
NM_001100407.2 |
661 |
Missense Mutation |
CTT,TTT |
L166F |
NP_001093877.1 |
NM_001100408.2 |
661 |
Missense Mutation |
CTT,TTT |
L152F |
NP_001093878.1 |
NM_001193653.1 |
661 |
Missense Mutation |
CTT,TTT |
L166F |
NP_001180582.1 |
NM_001193654.1 |
661 |
Missense Mutation |
CTT,TTT |
L166F |
NP_001180583.1 |
NM_001193655.1 |
661 |
Missense Mutation |
CTT,TTT |
L166F |
NP_001180584.1 |
NM_001193657.1 |
661 |
Missense Mutation |
CTT,TTT |
L166F |
NP_001180586.1 |
XM_006722293.2 |
661 |
Missense Mutation |
CTT,TTT |
L152F |
XP_006722356.1 |
XM_011523606.2 |
661 |
Missense Mutation |
CTT,TTT |
L199F |
XP_011521908.1 |
XM_017025072.1 |
661 |
Missense Mutation |
CTT,TTT |
L199F |
XP_016880561.1 |
XM_017025073.1 |
661 |
Missense Mutation |
CTT,TTT |
L185F |
XP_016880562.1 |
XM_017025074.1 |
661 |
Missense Mutation |
CTT,TTT |
L166F |
XP_016880563.1 |
XM_017025075.1 |
661 |
Missense Mutation |
CTT,TTT |
L166F |
XP_016880564.1 |
XM_017025076.1 |
661 |
Missense Mutation |
CTT,TTT |
L166F |
XP_016880565.1 |
XM_017025077.1 |
661 |
Missense Mutation |
CTT,TTT |
L166F |
XP_016880566.1 |
XM_017025078.1 |
661 |
Missense Mutation |
CTT,TTT |
L152F |
XP_016880567.1 |
- Gene
- HEXDC
- Gene Name
- hexosaminidase D
There are no transcripts associated with this gene.
View Full Product Details