Product Details
- SNP ID
-
rs11557539
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:83211162 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCTGTGCTGAACTGAAGGTGCCAAC[C/T]TCGGGACAGGGGCCACAGTCCACAA
- Phenotype
-
MIM: 154250
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ME1
PubMed Links
Gene Details
- Gene
- ME1
- Gene Name
- malic enzyme 1
View Full Product Details