Product Details

SNP ID
rs11554362
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:29767517 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCGAGCGCGCCTTCGATCAAGGCG[C/T]GGACGCTATCTACGACCACATCAAC
Phenotype
MIM: 610843
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UQCR10 PubMed Links

Gene Details

Gene
UQCR10
Gene Name
ubiquinol-cytochrome c reductase, complex III subunit X
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003684.1 149 Missense Mutation GCG,GTG A40V NP_001003684.1
NM_013387.3 149 Missense Mutation GCG,GTG A40V NP_037519.2
Gene
ZMAT5
Gene Name
zinc finger matrin-type 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003692.1 149 Intron NP_001003692.1
NM_001318129.1 149 Intron NP_001305058.1
NM_019103.2 149 Intron NP_061976.1

View Full Product Details