Product Details

SNP ID
rs11551584
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:97717973 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACTACTCTGGGCAGCTCCCATCC[A/C]AGTCTGGGGCCATTTGCAGACTCAG
Phenotype
MIM: 616970
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MARVELD1 PubMed Links

Gene Details

Gene
MARVELD1
Gene Name
MARVEL domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031484.3 3058 UTR 3 NP_113672.1

View Full Product Details