Product Details

SNP ID
rs11558068
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49185949 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAACCAGTTCCCCCACCAAAGCT[G/A]TGGAAAACCAAGAAGCCCTGGAGAC
Phenotype
MIM: 602529
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
TUBA1A PubMed Links

Gene Details

Gene
TUBA1A
Gene Name
tubulin alpha 1a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270399.1 850 Silent Mutation CAC,CAT H139H NP_001257328.1
NM_001270400.1 850 Silent Mutation CAC,CAT H104H NP_001257329.1
NM_006009.3 850 Silent Mutation CAC,CAT H139H NP_006000.2

View Full Product Details