Product Details

SNP ID
hCV176000322
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66271921 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCTGGGGTCCTGACTGGCAGCTG[G/T]GGGGAGAAGGGACAACACAGAGCAC
Phenotype
MIM: 611288 MIM: 611729 MIM: 612565
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CNIH2 PubMed Links

Gene Details

Gene
CNIH2
Gene Name
cornichon family AMPA receptor auxiliary protein 2
There are no transcripts associated with this gene.

Gene
KLC2
Gene Name
kinesin light chain 2
There are no transcripts associated with this gene.

Gene
RAB1B
Gene Name
RAB1B, member RAS oncogene family
There are no transcripts associated with this gene.

View Full Product Details