Product Details
- SNP ID
-
rs181109100
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:124611705 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTGTGGGCTCCCCCTTGTTGCTGA[A/T]GTCCCTGGCATTCGCCCAGGAGTTC
- Phenotype
-
MIM: 608422
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PANX3
PubMed Links
Gene Details
- Gene
- PANX3
- Gene Name
- pannexin 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_052959.2 |
149 |
Missense Mutation |
AAG,ATG |
K50M |
NP_443191.1 |
View Full Product Details