Product Details
- SNP ID
-
rs181199485
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:30378993 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCGCTGCTGGAAGATGGCGAGCGGC[A/C]GGGACGAGCGGCCGCCTTGGCGGCT
- Phenotype
-
MIM: 603102
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CPD
PubMed Links
Gene Details
- Gene
- CPD
- Gene Name
- carboxypeptidase D
View Full Product Details