Product Details

SNP ID
rs181126368
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53051866 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTCCACCTTCAGCCTAGGAAAGC[G/T]GAGCCTCATAGCTTCCGGGAGAAGG
Phenotype
MIM: 603928 MIM: 607717
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EIF4B PubMed Links

Gene Details

Gene
EIF4B
Gene Name
eukaryotic translation initiation factor 4B
There are no transcripts associated with this gene.

Gene
LOC283335
Gene Name
uncharacterized LOC283335
There are no transcripts associated with this gene.

Gene
MIR6757
Gene Name
microRNA 6757
There are no transcripts associated with this gene.

Gene
TNS2
Gene Name
tensin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015319.2 113 Silent Mutation GCG,GCT A39A NP_056134.2
NM_170754.2 113 Silent Mutation GCG,GCT A29A NP_736610.2
NM_198316.1 113 UTR 5 NP_938072.1
XM_006719302.3 113 Silent Mutation GCG,GCT A39A XP_006719365.1
XM_006719303.1 113 Silent Mutation GCG,GCT A29A XP_006719366.1
XM_011538079.1 113 Silent Mutation GCG,GCT A23A XP_011536381.1
XM_017019088.1 113 Silent Mutation GCG,GCT A29A XP_016874577.1
XM_017019089.1 113 Silent Mutation GCG,GCT A23A XP_016874578.1

View Full Product Details