Product Details

SNP ID
rs180867503
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:124963249 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTACCTGGCGCCCGAGCCGTTCTC[C/T]ATGCCCAGCCTGTTGGGAGCTCCAC
Phenotype
MIM: 602532
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNF217 PubMed Links

Gene Details

Gene
RNF217
Gene Name
ring finger protein 217
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286398.2 840 Silent Mutation TCC,TCT S235S NP_001273327.1
NM_152553.4 840 Intron NP_689766.1
XM_011535494.2 840 Silent Mutation TCC,TCT S235S XP_011533796.1
XM_011535495.2 840 Silent Mutation TCC,TCT S235S XP_011533797.1
XM_011535496.2 840 Silent Mutation TCC,TCT S235S XP_011533798.1
XM_011535499.2 840 Silent Mutation TCC,TCT S235S XP_011533801.1
XM_011535500.2 840 Intron XP_011533802.1
XM_017010317.1 840 Silent Mutation TCC,TCT S235S XP_016865806.1
Gene
RNF217-AS1
Gene Name
RNF217 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details