Product Details

SNP ID
rs180925087
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:160677632 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTGTGGACAGTACAGACAAACAG[C/T]GACTGGAAGAGTCTCAGAGACAGTT
Phenotype
MIM: 614439
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARL14 PubMed Links

Gene Details

Gene
ARL14
Gene Name
ADP ribosylation factor like GTPase 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025047.2 473 Silent Mutation CGA,TGA R96* NP_079323.1

View Full Product Details