Product Details

SNP ID
rs180953673
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:126256817 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGATCTTCCAGAATATTTCAAGC[A/G]ACTGACATGTGAGATCTCCACAAGG
Phenotype
MIM: 616587 MIM: 182180
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM118B PubMed Links

Gene Details

Gene
FAM118B
Gene Name
family with sequence similarity 118 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024556.3 2061 Missense Mutation CAA,CGA Q316R NP_078832.1
XM_005271665.3 2061 Missense Mutation CAA,CGA Q316R XP_005271722.1
XM_011542977.2 2061 Missense Mutation CAA,CGA Q316R XP_011541279.1
XM_011542978.2 2061 Missense Mutation CAA,CGA Q161R XP_011541280.1
XM_017018285.1 2061 Missense Mutation CAA,CGA Q316R XP_016873774.1
XM_017018286.1 2061 Missense Mutation CAA,CGA Q161R XP_016873775.1
Gene
SRPRA
Gene Name
SRP receptor alpha subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177842.1 2061 Intron NP_001171313.1
NM_003139.3 2061 Intron NP_003130.2
XM_017018179.1 2061 UTR 3 XP_016873668.1

View Full Product Details