Product Details

SNP ID
rs180935964
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:65518087 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGACTCTTGCCCTGTAGAGAAAGG[C/T]CCCACCTCCATTCTGTCTTCCTCTT
Phenotype
MIM: 300964
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LAS1L PubMed Links

Gene Details

Gene
LAS1L
Gene Name
LAS1 like, ribosome biogenesis factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170649.1 1852 Silent Mutation GGA,GGG G592G NP_001164120.1
NM_001170650.1 1852 Silent Mutation GGA,GGG G550G NP_001164121.1
NM_031206.4 1852 Silent Mutation GGA,GGG G609G NP_112483.1
XM_005262301.1 1852 Silent Mutation GGA,GGG G609G XP_005262358.1
XM_005262304.1 1852 Intron XP_005262361.1
XM_005262305.3 1852 Intron XP_005262362.1
XM_005262306.3 1852 Intron XP_005262363.1
XM_011531045.1 1852 Silent Mutation GGA,GGG G567G XP_011529347.1
XM_011531046.1 1852 Intron XP_011529348.1
XM_017029877.1 1852 Silent Mutation GGA,GGG G592G XP_016885366.1
XM_017029878.1 1852 Intron XP_016885367.1
XM_017029879.1 1852 Silent Mutation GGA,GGG G307G XP_016885368.1
XM_017029880.1 1852 Silent Mutation GGA,GGG G290G XP_016885369.1

View Full Product Details