Product Details

SNP ID
rs181001143
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:149844688 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGAGGAATGGGTGCTGCTGAGC[C/T]AGCAACAGAAGGAGCTCTGTGGTTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF862 PubMed Links

Gene Details

Gene
ZNF862
Gene Name
zinc finger protein 862
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099220.1 290 Nonsense Mutation CAG,TAG Q30* NP_001092690.1
XM_006716085.3 290 Nonsense Mutation CAG,TAG Q44* XP_006716148.1
XM_011516463.1 290 Nonsense Mutation CAG,TAG Q44* XP_011514765.1
XM_011516464.1 290 Nonsense Mutation CAG,TAG Q37* XP_011514766.1
XM_011516465.2 290 Intron XP_011514767.1
XM_011516466.2 290 Intron XP_011514768.1
XM_017012519.1 290 Nonsense Mutation CAG,TAG Q23* XP_016868008.1

View Full Product Details